ENA313839441 Enamine

1-[(3,4-dichlorophenyl)methyl]piperidin-4-amine

  • MDL number MFCD10005787
  • Cas Number : 92539-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73179
Availability details for
Quantity
  • Availability
  • Add to Cart