ENA313839444 Enamine

prop-2-en-1-yl({1-[3-(1H-1,2,3,4-tetrazol-1-yl)phenyl]ethyl})amine

  • MDL number MFCD11138112
  • Cas Number : 1019612-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73183
Availability details for
Quantity
  • Availability
  • Add to Cart