ENA313839447 Enamine

2-chloro-N-[1-(2,6-difluorophenyl)propan-2-yl]acetamide

  • MDL number MFCD12798917
  • Cas Number : 1179059-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-47882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47882
Availability details for
Quantity
  • Availability
  • Add to Cart