ENA313839453 Enamine

1-(4-methylpiperazin-1-yl)-2-(piperidin-4-yl)ethan-1-one

  • MDL number MFCD02684229
  • Cas Number : 397869-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73208

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73208
Availability details for
Quantity
  • Availability
  • Add to Cart