ENA313839455 Enamine

methyl[(4-methylphenyl)(pyridin-2-yl)methyl]amine

  • MDL number MFCD12154227
  • Cas Number : 1178242-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73210
Availability details for
Quantity
  • Availability
  • Add to Cart