ENA313839461 Enamine

5-(2-phenylethyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11156015
  • Cas Number : 192505-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72431

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72431
Availability details for
Quantity
  • Availability
  • Add to Cart