ENA313839462 Enamine

1-{1-[(4-methylbenzene)sulfonyl]-1H-pyrrol-3-yl}ethan-1-one

  • MDL number MFCD00143087
  • Cas Number : 106058-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73220
Availability details for
Quantity
  • Availability
  • Add to Cart