ENA313839474 Enamine

2-[(piperidine-1-sulfonyl)amino]ethan-1-ol

  • MDL number MFCD12185468
  • Cas Number : 181762-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73239
Availability details for
Quantity
  • Availability
  • Add to Cart