ENA313839479 Enamine

6-tert-butyl-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carbonitrile

  • MDL number MFCD16437743
  • Cas Number : 1268129-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73261
Availability details for
Quantity
  • Availability
  • Add to Cart