ENA313839481 Enamine

(1S)-1-(4-iodophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD16295044
  • Cas Number : 1308650-40-9
  • Molecular Weight : 283.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73263
Availability details for
Quantity
  • Availability
  • Add to Cart