ENA313839485 Enamine

phenyl N-(1H-pyrazol-4-yl)carbamate

  • MDL number MFCD12133963
  • Cas Number : 1154876-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72513
Availability details for
Quantity
  • Availability
  • Add to Cart