ENA313839494 Enamine

methyl 4-methanesulfinylbutanoate

  • MDL number MFCD14613376
  • Cas Number : 1249193-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73281
Availability details for
Quantity
  • Availability
  • Add to Cart