ENA313839497 Enamine

3-bromo-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD02135252
  • Cas Number : 23095-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73295
Availability details for
Quantity
  • Availability
  • Add to Cart