ENA313839498 Enamine

2-{[4-(aminomethyl)-2-fluorophenyl]methoxy}ethan-1-ol

  • MDL number MFCD13808635
  • Cas Number : 1019546-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73296
Availability details for
Quantity
  • Availability
  • Add to Cart