ENA313839517 Enamine

1-(3-aminophenyl)-3-(5,6,7,8-tetrahydronaphthalen-1-yl)urea

  • MDL number MFCD13374525
  • Cas Number : 1040346-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73323
Availability details for
Quantity
  • Availability
  • Add to Cart