ENA313839533 Enamine

6-methyl-2-(4-phenylphenyl)pyrimidin-4-ol

  • MDL number MFCD18380738
  • Cas Number : 1154551-68-4
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73368
Availability details for
Quantity
  • Availability
  • Add to Cart