ENA313839534 Enamine

2,3-dihydro-1,4-benzodioxine-6-carbonitrile

  • MDL number MFCD00221462
  • Cas Number : 19102-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73369
Availability details for
Quantity
  • Availability
  • Add to Cart