ENA313839536 Enamine

[(2-ethyl-1,3-thiazol-5-yl)methyl](methyl)amine

  • MDL number MFCD02853612
  • Cas Number : 1184195-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73372
Availability details for
Quantity
  • Availability
  • Add to Cart