ENA313839542 Enamine

3-amino-N-(5-chloropyridin-2-yl)benzamide

  • MDL number MFCD11184857
  • Cas Number : 1039827-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73383
Availability details for
Quantity
  • Availability
  • Add to Cart