ENA313839543 Enamine

2-chloro-N-{1-[(methylcarbamoyl)methyl]-1H-pyrazol-4-yl}acetamide

  • MDL number MFCD12798946
  • Cas Number : 1178226-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-48861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48861
Availability details for
Quantity
  • Availability
  • Add to Cart