ENA313839546 Enamine

diethyl[3-(methylamino)propyl]amine

  • MDL number MFCD00026748
  • Cas Number : 5459-95-0
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73388
Availability details for
Quantity
  • Availability
  • Add to Cart