ENA313839549 Enamine

1-(3-bromophenyl)pyrrolidin-3-one

  • MDL number MFCD11640418
  • Cas Number : 1096881-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73391

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73391
Availability details for
Quantity
  • Availability
  • Add to Cart