ENA313839557 Enamine

2-(furan-2-amido)thiophene-3-carboxylic acid

  • MDL number MFCD09046215
  • Cas Number : 926190-41-2
  • Molecular Weight : 237.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73408
Availability details for
Quantity
  • Availability
  • Add to Cart