ENA313839560 Enamine

6-(2,6-difluorophenyl)pyridazin-3-amine

  • MDL number MFCD16860046
  • Cas Number : 1250246-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73412

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-73412
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart