ENA313839564 Enamine

[1-(cyclohexylmethyl)pyrrolidin-2-yl]methanamine

  • MDL number MFCD10003504
  • Cas Number : 72135-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73418
Availability details for
Quantity
  • Availability
  • Add to Cart