ENA313839567 Enamine

2-(chloromethyl)-5-(thiophen-2-yl)-1,3-oxazole

  • MDL number MFCD09880832
  • Cas Number : 70996-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73422
Availability details for
Quantity
  • Availability
  • Add to Cart