ENA313839569 Enamine

1-[ethyl(2-methylphenyl)amino]-3-phenylpropan-2-one

  • MDL number MFCD13802490
  • Cas Number : 1157615-03-6
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73425
Availability details for
Quantity
  • Availability
  • Add to Cart