ENA313839572 Enamine

{1-[(2-methyl-1,3-thiazol-4-yl)methyl]piperidin-4-yl}methanamine

  • MDL number MFCD16110195
  • Cas Number : 1249955-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73430

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73430
Availability details for
Quantity
  • Availability
  • Add to Cart