ENA313839574 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD12540701
  • Cas Number : 1097017-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73432

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73432
Availability details for
Quantity
  • Availability
  • Add to Cart