ENA313839578 Enamine

3-chloro-4-(difluoromethoxy)benzaldehyde

  • MDL number MFCD16075291
  • Cas Number : 1121585-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73437
Availability details for
Quantity
  • Availability
  • Add to Cart