ENA313839590 Enamine

1-[(4-bromo-2-fluorophenyl)methyl]-4-methyl-1H-pyrazole

  • MDL number MFCD14686899
  • Cas Number : 1248326-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73469
Availability details for
Quantity
  • Availability
  • Add to Cart