ENA313839592 Enamine

methyl 2-(3-methylphenoxy)propanoate

  • MDL number MFCD12031801
  • Cas Number : 125095-33-2
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73471
Availability details for
Quantity
  • Availability
  • Add to Cart