ENA313839595 Enamine

3-(4-fluorophenyl)-1-(1,2,3,4-tetrahydroisoquinolin-5-yl)urea

  • MDL number MFCD13806051
  • Cas Number : 1157922-50-3
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73474
Availability details for
Quantity
  • Availability
  • Add to Cart