ENA313839598 Enamine

8-azabicyclo[3.2.1]octan-3-ol

  • MDL number MFCD16664490

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51588

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51588
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart