ENA313839599 Enamine

1-(3-nitrophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10696381
  • Cas Number : 944905-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73480
Availability details for
Quantity
  • Availability
  • Add to Cart