ENA313839601 Enamine

tert-butyl N-(3,5-dimethyl-1H-pyrazol-4-yl)carbamate

  • MDL number MFCD12135414
  • Cas Number : 1187341-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73484
Availability details for
Quantity
  • Availability
  • Add to Cart