ENA313839603 Enamine

5-[(2-chlorophenyl)methyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09803628
  • Cas Number : 862574-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72601
Availability details for
Quantity
  • Availability
  • Add to Cart