ENA313839604 Enamine

6,7-difluoro-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD16745183
  • Cas Number : 137690-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279-280
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73487
Availability details for
Quantity
  • Availability
  • Add to Cart