ENA313839605 Enamine

3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine hydrochloride

  • MDL number MFCD11110675
  • Cas Number : 733757-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73489
Availability details for
Quantity
  • Availability
  • Add to Cart