ENA313839607 Enamine

1,2,4-oxadiazol-3-ylmethanamine hydrochloride

  • MDL number MFCD16990589
  • Cas Number : 370103-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73491

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-73491
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart