ENA313839609 Enamine

1-[(2S)-2-ethynylpiperidin-1-yl]ethan-1-one

  • MDL number MFCD09263910
  • Cas Number : 128960-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73494
Availability details for
Quantity
  • Availability
  • Add to Cart