ENA313839616 Enamine

2-(cyanomethoxy)-5-methoxybenzoic acid

  • MDL number MFCD11618477
  • Cas Number : 1094687-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73516
Availability details for
Quantity
  • Availability
  • Add to Cart