ENA313839617 Enamine

5-(2-phenoxyethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09816920
  • Cas Number : 1016680-14-0
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72603
Availability details for
Quantity
  • Availability
  • Add to Cart