ENA313839626 Enamine

6-(thiophen-2-yl)hexanoic acid

  • MDL number MFCD12172599
  • Cas Number : 21010-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73536
Availability details for
Quantity
  • Availability
  • Add to Cart