ENA313839628 Enamine

6-(2-methylpropanoyl)-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD11211452
  • Cas Number : 70735-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73539
Availability details for
Quantity
  • Availability
  • Add to Cart