ENA313839629 Enamine

3-(3-methylphenyl)-1-(1,2,3,4-tetrahydroquinolin-8-yl)urea

  • MDL number MFCD09804030
  • Cas Number : 1019116-11-0
  • Molecular Weight : 281.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73540
Availability details for
Quantity
  • Availability
  • Add to Cart