ENA313839640 Enamine

3-amino-1-phenylpyrrolidin-2-one

  • MDL number MFCD14689622
  • Cas Number : 1904-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73584
Availability details for
Quantity
  • Availability
  • Add to Cart