ENA313839645 Enamine

cyclopentyl(2-methoxyphenyl)methanone

  • MDL number MFCD03841294
  • Cas Number : 7063-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73593
Availability details for
Quantity
  • Availability
  • Add to Cart