ENA313839648 Enamine

N-(cyclopropylmethyl)-1H-1,3-benzodiazole-5-carboxamide

  • MDL number MFCD12194504
  • Cas Number : 1156039-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73597
Availability details for
Quantity
  • Availability
  • Add to Cart