ENA313839650 Enamine

4-(3-aminopiperidin-1-yl)-3-fluorobenzonitrile

  • MDL number MFCD14617970
  • Cas Number : 1248383-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73599
Availability details for
Quantity
  • Availability
  • Add to Cart